Thermo Scientific™ GeneJET™ PCR Purification Kit PROMO
Perform rapid and efficient purification of DNA from PCR and other enzymatic reaction mixtures with this kit.
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: K0702-RMP6225
UNSPSC: 41105504
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 15503531
Quantity Price
1 £ 251.0 / Each
Estimated Shipment date
from Supplier 03-03-2017
Add to basket
Description and Specification
Highlights
  • Highly efficient: 90 to 100% recoveries in the range of 100 bp to 10 kb
  • Fast: procedure takes only 5 minutes
  • Convenient: spin columns are capped and assembled with collection tubes