Thermo Scientific™ GeneJET™ Gel Extraction Kit PROMO
Perform silica membrane-based DNA fragment purification from standard or low-melting point agarose gels run in either TAE or TBE buffer.
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: K0691-RMP6225
X1 GeneJET Gel Extraction Kit
UNSPSC: 41105501
Additional Details:
Additional Details: Weight: 0.35000kg
Product Code. 15573521
Quantity Price
1 £ 61.5 / Each
EU Stock 523
Add to basket
Description and Specification
Highlights
  • Highly efficient: up to 95% recoveries in the range of 100 bp to 10 kb
  • Fast: procedure takes only 15 minutes
  • Convenient: spin columns are capped and assembled with collection tubes