missing translation for 'onlineSavingsMsg'
Learn More

Gelatine, Extra Pure, SLR, Granular Powder, meets analytical specification of Ph.Eur., Fisher Chemical

£30.68 - £64.15

Chemical Identifiers

CAS 9000-70-8
MDL Number 81638
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11442624 SDS View Documents Fisher Chemical
G/0150/50
250g £30.68
250g View alternative products
Save Up to 
»
In Stock 7
Please to purchase this item. Need a web account? Register with us today!
10774751 SDS View Documents Fisher Chemical
G/0150/53
500g £64.15
500g
Qty Price Break Available View alternative products
Save Up to 
»
In Stock 37
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

9000-70-8
81638

Specifications

Plastic powder jar
250g
9000-70-8
Powder Solid
Beige
5 to 7
81638