Learn More
Sartorius™ Gelatin Filter Disposables
Brand:  Sartorius™ 17528-080BZD
UNSPSC : 41104008
Code : 04
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11556644
  • £145.05 / Pack of 10
Estimated Shipment
29-06-2017
Add to basket
Specifications
Description
  • Ideal for the pharmaceutical, biotechnology, food and beverage industries and hospitals
  • Accu-powered and portable
  • Accu-control guarantees constant performance during air sampling
  • Ergonomic and easy to clean
  • Flexible adjustment of flow and sample volume
  • Gelatin membrane filter method guarantees exact measurements
Description & Specifications
Specifications
Gelatin Filter Disposables
10Pack
Documentation