Learn More
Sartorius™ Gelatin Filter Disposables
Brand:  Sartorius™ 17528-080ACD
UNSPSC : 41104008
Code : 53
Additional Details : Weight : 0.01100kg
Product Code. 11546644
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Description
  • Ideal for the pharmaceutical, biotechnology, food and beverage industries and hospitals
  • Accu-powered and portable
  • Accu-control guarantees constant performance during air sampling
  • Ergonomic and easy to clean
  • Flexible adjustment of flow and sample volume
  • Gelatin membrane filter method guarantees exact measurements
Description & Specifications
Specifications
10Pack
Gelatin Filter Disposables
Documentation Alternative Products