missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gelatin Blocking Buffer, 1% in TBS

Brand:  Alfa Aesar™ J62966.K2

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15435259

  • £55.65 / 1 L


Estimated Shipment date
from Supplier 10-07-2017
Add to basket

Specifications Description & Specifications

Specifications

Blocking Buffer
1% in TBS
Liquid
1L
Documentation

Technical Documents