missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Alfa Aesar Gelatin Blocking Buffer, 1% in TBS, with 0.02% sodium azide
Brand: Thermo Scientific Alfa Aesar J60805.AP
Gelatin Blocking Buffer, 1% in TBS, with 0.02% sodium azide
Code : NEW
Additional Details : Weight : 0.00050kg