missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gelatin Blocking Buffer, 1% in TBS, with 0.02% sodium azide

Brand:  Alfa Aesar™ J60805.K2

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15443509

  • £55.65 / 1 L


Estimated Shipment date
from Supplier 10-07-2017
Add to basket

Specifications Description & Specifications

Specifications

1L
Blocking Buffer
Liquid
Documentation

Technical Documents