missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gelatin Blocking Buffer, 1% in PBS, with 0.02% sodium azide

Brand:  Alfa Aesar™ J63104.K2

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15415379

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Liquid
Blocking Buffer
1L
Videos
SDS