missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for Easigel H1-SET Horizontal Gel Electrophoresis Unit

For use with Easigel H1-SET Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11700555

  • £15.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Color coded
  • Offer flexibility in sample-loading
  • Volume to maximum 40-sample throughput
Specifications

Specifications

1.5mm
Easigel H1-SET Horizontal Gel Electrophoresis Unit
27μL
Comb
16
4mm
Videos
Documentation

Technical Documents