missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for HU20 Maxi-Standard Horizontal Gel Electrophoresis Unit, 1mm thick

For use with HU20 Maxi-Standard Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 2.00000kg

Product Code. 10364971

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Color coded
  • Height adjustable
  • Offer complete control over loading volume and well depth
Specifications

Specifications

1mm
HU20 Maxi-Standard Horizontal Gel Electrophoresis Unit
20μL
Comb
28
5mm
Videos
Documentation

Technical Documents