missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for HU20 Maxi-Standard Horizontal Gel Electrophoresis Unit, 1mm thick

For use with HU20 Maxi-Standard Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 2.00000kg

Product Code. 10548441

  • £28.00 / Each


Estimated Shipment
18-07-2018
Add to basket

Description

Description

  • Color coded
  • Height adjustable
  • Offer complete control over loading volume and well depth
Specifications

Specifications

1mm
HU20 Maxi-Standard Horizontal Gel Electrophoresis Unit
35μL
Comb
16
8.5mm
Videos
Documentation

Technical Documents