missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for Easigel H1-SET Horizontal Gel Electrophoresis Unit
For use with Easigel H1-SET Horizontal Gel Electrophoresis Unit
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.80000kg
Description
- Color coded
- Offer flexibility in sample-loading
- Volume to maximum 40-sample throughput
Specifications
Easigel H1-SET Horizontal Gel Electrophoresis Unit | |
12 | |
27 μL |
Comb | |
1 mm | |
6 mm |