missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for HU13 Midi Horizontal Gel Electrophoresis Unit, 1.5mm thick

For use with HU13 Midi Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 10491083

  • £27.00 / Each


Estimated Shipment
25-07-2018
Add to basket

Description

Description

  • Color coded
  • Height adjustable
  • Offer complete control over loading volume
  • Well depth to maximum 112-sample throughput
Specifications

Specifications

1.5mm
HU13 Midi Horizontal Gel Electrophoresis Unit
20μL
Comb
28
3mm
Videos
Documentation

Technical Documents