missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for HU6 Mini Horizontal Gel Electrophoresis Unit

For use with HU6 Mini Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 2.00000kg

Product Code. 10233321

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Color coded
  • Height adjustable
  • Offer complete control over loading volume
  • Well depth to maximum 32-sample throughput
Specifications

Specifications

1mm
HU6 Mini Horizontal Gel Electrophoresis Unit
10μL
Comb
16
2.2mm
Videos
Documentation

Technical Documents