missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for VG-SYS Vari-gel MAXI Horizontal Electrophoresis Unit
For use with VG-SYS Vari-gel MAXI Horizontal Electrophoresis Unit.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.80000kg
Description
- Color coded
- For use with VG-SYS Vari-gel MAXI Horizontal Electrophoresis Unit
Specifications
VG-SYS Vari-Gel MAXI Horizontal Electrophoresis Unit | |
10 | |
2 mm | |
12.8 mm |
Comb | |
1 Each | |
128 μL |