missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for HU10 Mini-Plus Horizontal Gel Electrophoresis Unit, 2mm thick

For use with HU10 Mini-Plus Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.17000kg

Product Code. 10233181

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Color coded
  • Height adjustable
  • Offer complete control over loading volume and well depth to maximum 32-sample throughput
Specifications

Specifications

2mm
HU10 Mini-Plus Horizontal Gel Electrophoresis Unit
25μL
Comb
20
3mm
Videos
Documentation

Technical Documents