missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for HU13 Midi Horizontal Gel Electrophoresis Unit, 2mm thick

For use with HU13 Midi Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01000kg

Product Code. 12368426

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Color coded
  • Height adjustable
  • Offer complete control over loading volume
  • Well depth to maximum 112-sample throughput
Specifications

Specifications

2mm
HU13 Midi Horizontal Gel Electrophoresis Unit
35μL
Comb
20
4mm
Alternative Products