missing translation for 'onlineSavingsMsg'
Learn More

Ultrospec™ 2100 pro UV/Visible Spectrophotometers

Simple-to-use instrument for nucleic acid measurements and enzyme kinetics

Brand:  Harvard Bioscience 80211221

Code : GE

Additional Details : Weight : 19.66250kg

 View more versions of this product

Product Code. 10477864

  • £5210.00 / Each


Estimated Shipment
02-08-2018
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Specifications

Specifications

±0.5% or ±0.003A to 3.000A at 54nm
14cm
100 to 240V 50/60Hz
8cm
Xenon
>3nm
±1nm
13kg
3 Year
CE89/336/EEC (EMCdirective); CE73/23/EEC (LVdirective); EN-61010-1 (IEC1010-1)
Two Silicon Photodiodes
Nucleic acid measurements and enzyme kinetics
50/60Hz
3000nm/min.
100 to 240V
190 to 900nm
21cm
UV-visible
Videos