missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Tricorn™ Superose™ 6 and Superose 12 Prepacked Gel Filtration Columns

Designed for high performance, laboratory scale separations of proteins and other biomolecules based on size.  GE Healthcare Tricorn™ Superose™ 6 and Superose 12 Prepacked Gel Filtration Columns  delivers quick and reproducible separations with high resolution.

Brand:  GE Healthcare 17-5172-01

Code : GX

Additional Details : Weight : 0.99000kg

 View more versions of this product

Product Code. 10014647

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

 

  • Covers wide separation range of molecular weights from 1000 up to 5million MHighly cross-linked agarose matrixSmall average bead size of 13μm (Superose 6) and 11μm (Superose 12) are designed for laboratory-scale separations with good resolving capability
  • Durable, high-performance glass columns for reliable and reproducible separations
  • Available with two different media - Superose 6 or Superose 12

     

     

  • Bed volume: 24mL
  • Bed dimension: 10 × 300mm
  • Pressure max.: 15 bar (217psi)

  • Media

  • pH stability working range: 3 to 12
  • pH stability cleaning: 1 to 14

  • Column

  • Column I.D.: 10mm
  • Bed support: filter
  • Material (bed support): polypropylene
  • Material (column hardware): borosilicate glass, PEEK, PP, EPDM and PFR
  • Material (column tube): borosilicate glass
  • Bed height with one adaptor: 279 to 309mm
  • Bed height with two adaptors: 245 to 306mm
  •  

    Superose 6 Media:

  • Average partcle size: 13μm
  • Gel filtration separations over a very broad molecular weight rang from 5000 to 5,000,000 (globular proteins)
  • Flow rate: 0.3 to 0.5mL/min.

  • Superose 12 Media:

  • Average partcle size: 11μm
  • Gel filtration separations over a very broad molecular weight rang from 1000 to 300,000 (globular proteins)
  • Flow rate of 0.5 to 1mL/min.
  •  

Specifications

Specifications

1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 1% SDS, Organic Solvents, 0.5M NaOH (for cleaning in-place)
10mm
Each
300mm
Valco fittings for AKTA™ design systems and are supplied with M6 connectors for FPLC System
217psi
13μm
4° to 30°C
24mL
Superose 6 prepacked column
High performance gel filtration with a broad protein fractionation range of 5000 to 5,000,000
0.3 to 0.5mL/min.
Borosilicate Glass
3 to 12
Cross-linked Agarose
Videos