missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Tricorn™ Superdex™ Prepacked Gel Filtration Columns

Use for laboratory-scale separations and intermediate purification based on different sizes of proteins, peptides, and other biomolecules. GE Healthcare Life Sciences™Tricorn™ Superdex™ Prepacked Gel Filtration Columns are perfect for high-resolution applications.

Brand:  GE Healthcare 17-5176-01

Code : GX

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10729134

  • £1593.38 / Each


Estimated Shipment
09-02-2018
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

  • Fast gel filtration involving separation based on three molecular weight ranges from 100 to 600000 M
  • Spherical composite of cross-linked agarose and dextran matrix
  • Comes with two different bed heights of 150 and 300mm allowing greater flexibility to suit different analytical needs
  • Available with three different media: Superdex Peptide, Superdex 75, and Superdex 200

Superdex Peptide:

  • Bed dimension: 10 x 300mm
  • Bed volume: 24mL
  • Flow velocity: 0.15 to 0.6mL/min.
  • Pressure max.: 18 bar (261psi)
  • Separation range: 100 to 7000 (peptides)
  • pH Stability working range: 1 to 14
  • Column I.D.: 10mm
  • Bed height with one adaptor:     279 to 309mm
  • Bed height with two adaptors: 245 to 306mm
Superdex 200:
  • Bed dimension: 5 x 150mm and 10 x 300mm
  • Bed volume: 3 and 24mL
  • Flow velocity: 0.15 to 0.6mL/min. and 0.25 to 0.75mL/min.
  • Pressure max.: 15 bar (217psi)
  • Separation range: 10,000 to 600,000 (globular peptides)
  • pH Stability working range: 3 to 12
  • Column I.D.: 5 and 10mm
  • Bed height with one adaptor:     135 to 159mm (3mL column)
  • Bed height with two adaptors: 108 to 156mm (3mL column)
Superdex 75:
  • Bed dimension: 5 x 150mm and 10 x 300mm
  • Bed volume: 3 and 24mL
  • Flow velocity: 0.15 to 0.6mL/min.
  • Pressure max.: 18 bar (261psi)
  • Separation range: 3000 up to 70,000 (globular proteins)
  • pH Stability working range: 3 to 12
  • Column I.D.: 5 and 10 mm
  • Bed height with one adaptor: 108 to 156mm (3mL column)
  • Bed height with two adaptors: 135 to 159mm (3mL column)

Specifications

Specifications

1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol, 70% Ethanol, 1M NaOH
10mm
Each
300mm
Valco fittings for AKTA™ design systems and are supplied with M6 connecto rs for FPLC System
18 bar
13μm
4° to 30°C
24mL
Superdex™ Peptide prepacked column
High-resolution, semipreparative and analytical separations of biomolecules
0.15 to 0.6mL/min.
Borosilicate Glass
1 to 14
Spherical composite of cross-linked Agarose and Dextran
Videos