missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva StrepTactin™ Sepharose™ High Performance Medium
Mild purification conditions preserve antigenicity and function. Cytiva StrepTactin Sepharose High Performance media is used for robust, high resolution purification of Strep II-tagged proteins.
£552.60 - £2454.03
Specifications
Product Type | StrepTactin Sepharose HP media |
---|---|
Flow Rate | <300 cm/hr. |
Ligand Type | StrepTactin (∼5mg StrepTactin/mL medium) |
pH Range | 7 |
Particle Size | 34μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10762827
|
Cytiva
28-9356-00 |
50mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10636895
|
Cytiva
28-9355-99 |
10mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Medium with high binding capacity yields high resolution, sharp concentrated peaks, and reproducible purifications
- Physiological conditions and mild elution preserves the activity of the target protein
- Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
- Easily scaled up, and easily regenerated with 0.5M NaOH
- Convenient use with a peristaltic pump or chromatography system
- Matrix: rigid, highly cross-linked 6% agarose
- Ave. particle size: 34μm
- Dynamic binding capacity (protein dependent): approx. 6mg Strep(II)-tagged protein/mL medium
- Compatibility: stable in all commonly used buffers, 0.5M NaOH (regeneration and cleaning), reducing agents, denaturants, and detergents
- pH stability: >7
Specifications
StrepTactin Sepharose HP media | |
StrepTactin (∼5mg StrepTactin/mL medium) | |
34μm | |
2° to 8°C | |
Strep-Tactin Sepharose |
<300 cm/hr. | |
7 | |
6% Highly Cross-linked Agarose | |
Purification of Strep (II) -tagged recombinant Proteins to high purities in concentrated forms and small volumes by using with perstaltic pump or chromato graphy system |