missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Sephadex™ Ion Exchange Media

Separate molecules over broad molecular weight and pH ranges

Brand:  GE Healthcare 17-0240-01

Code : GE

Additional Details : Weight : 0.13800kg

 View more versions of this product

Product Code. 11564895

  • £271.49 / 100g


Call for latest availability
+44 (0)1509 555 500
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

Range of weak and strong anionic (A) and cationic (C) exchangers. Fractionation range: A-25 and C-25, up to 30 000; A-50 and C-50: 30 000–100 000.

Very high capacities even at elevated ionic strengths
Very low non-specific adsorption
Wide range of applications at laboratory and process scales
Suitable for column or batch techniques
Simple and economical to use

DEAE Sephadex

  • Weak and strong anion exchanger
  • pH range 2-9

QAE Sephadex
  • Strong anion exchanger
  • pH range: 2-12
CM Sephadex
  • Weak cation exchanger
  • pH range: 6-10
SP Sephadex
  • Strong cation exchanger
  • pH range: 4-13

Specifications

Specifications

100g
<45cm/hr.
Sephadex cation exchange media
Separating and purifying Proteins, Polypeptides, Nucleic Acids and other charged biomolecules by using column or batch techniques
Sulphopropyl
40 to 120μm in dry form
Cross-linked Dextran
4° to 25°C
Column or Batch Techniques
Solid Spherical Bead
5 to 15cm
SP-Sephadex C-50
2 to 12
30,000 to 100,000
Videos