GE Healthcare Sample Applicator for PhastSystem™ Electrophoresis System
Sample applicators for PhastSystem Electrophoresis Systems
Overview
Brand: GE Healthcare
Manufacturer Part Number: 18161801
X50 Sample applicators GE Healthcare Phastgel 8 x
UNSPSC: 41105305
Code: GE
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 10328364
Quantity Price
1 £ 114.71000000000001 / Pack of 50
Estimated Shipment
16-03-2017
Add to basket
If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.
Description and Specification
Specification
Product Type Sample Applicator
For Use With (Equipment) PhastGel™ system
Device Holds 8 x 1μL
Documentation
Technical documents