missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva rmp Protein A Sepharose™ Fast Flow Affinity Media
Designed for high-purity separation of monoclonal and polyclonal antibodies
£677.00 - £5280.15
Specifications
For Use With (Application) | For High-Purity Separation of Monoclonal and Polyclonal Antibodies |
---|---|
Stability | Stable in aqueous Buffers commonly used in Protein A Chromatography |
Storage Requirements | 2°C to 8°C |
Binding Capacity | >Min. 22 mg Human IgG/mL |
Product Type | rmp Protein A Sepharose Fast Flow Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11369981
|
Cytiva
17-5138-01 |
5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11349981
|
Cytiva
17-5138-02 |
25 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10358374
|
Cytiva
17-5138-03 |
200 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Non-mammalian-based media composed of recombinant Protein A coupled with Sepharose 4 Fast Flow base matrix by reductive amination
- Coupling chemistry that minimizes leakage of protein A ligand
- Multi-point attachment of rProtein A by chemically-stable amide bonds
- High capacity and good pressure/flow characteristics provide for high throughput
- Matrix: 4% cross-linked agarose
- Average particle size: 90μm (d50V)
- Ligand: recombinant protein A (E. coli)
- pH Stability working range: 3 to 10
- pH Stability cleaning-in-place: 2 to 11
- Binding capacity/mL chromatography medium: >35mg human IgG/mL
- Pressure/Flow specification: base matrix 150 to 250cm/hr., 100kPa, XK 50/60 column
For therapeutic applications requiring extremely pure eluate fractions of antibodies with non-exposure of antibodies to human or animal derivatives during purification process.
Specifications
For High-Purity Separation of Monoclonal and Polyclonal Antibodies | |
2°C to 8°C | |
rmp Protein A Sepharose Fast Flow Media | |
4% Cross-Linked Agarose | |
20% Ethanol |
Stable in aqueous Buffers commonly used in Protein A Chromatography | |
>Min. 22 mg Human IgG/mL | |
60 to 165 μm | |
3 to 10 | |
Recombinant Protein A (E. coli) |