Learn More
Cytiva RESOURCE™ and SOURCE™ 15 and 5RPC columns
High Performance Reversed Phase Chromatography
Brand: Cytiva 17-5068-01
Code : GE
Additional Details : Weight : 1.78720kg
Description
Ideally suited for difficult preparative separations at all scales
- High resolution separations in minutes
- High performance at low back pressures
- High capacity at high flow rates
- Wide pH stability range
- Excellent scalability
- Reproducible quality
Specifications
4.6 mm | |
100 mm | |
40 bar (580 psi) | |
15 μm | |
1 Pc. | |
Polystyrene/Divinyl Benzene | |
Stainless Steel (316) |
0.5 to 2.5 mL/min | |
Matrix surface | |
Monosized | |
1 to 12 | |
1.7 mL | |
No | |
Reversed Phase Column |
Safety and Handling
- SOURCE 15RPC, ST 4.6/100
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.