Learn More
GE Healthcare Recombinant Taq DNA Polymerase
For PCR and other procedures requiring DNA polymerase activity at elevated temperatures
Brand:  GE Healthcare 28-9373-45
Code : GX
Additional Details : Weight : 0.01400kg
 View more versions of this product
Product Code. 11339951
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
This product has been reformulated such that it does not fall under the remaining claims of US patents 5,079,352, 5,789,224, 5,618,711, 6,127,155 and their foreign equivalents outside Canada. Therefore no license to practise under the aforementioned patents is conveyed by purchase of this product. No other patent rights (such as 5' Nuclease Process patent rights in US Patents Nos. 5,210,015 and 5,487,972) are conveyed expressly, by implication, or by estoppel.
Specifications
Specifications
PCR and other procedures requiring DNA polymerase activity at elevated temperatures
E. coli
Taq DNA polymerase, 10x PCR buffer
1000U
Liquid
7.5
Recombinant Taq DNA Polymerase
Videos