missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Processor Plus Blot Processing Trays
For blot processing
£182.58 - £1961.46
Specifications
Temperature (English) Operating | 39.2°F to 104°F |
---|---|
Temperature (Metric) Operating | 4°C to 40°C |
Includes: For Blot Processing Tray Pack
- One tray support
- Three blot processing mini trays
- Three blot processing standard trays
- One blot lid assembly with valves and tubing
- One blot tray base
- Four reagent bottles and a rack
- Two wast bottle caps
- One spirit level
- One lid support
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
10476724
|
Cytiva
80-6444-23 |
Processor Plus™ System | Blot Processing Tray Pack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10350845
|
Cytiva
80-6444-42 |
Processor Plus™ System for Gel Staining and Blot Processing | Blot Processing Mini Tray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Blot Processing Mini Tray
- Tray volume: 10 to 50mL per chamber
- Operating temperature: 4° to 40°C
- Trays for four mini- (up to 9 × 9.5cm) standard sized blots
- High-speed peristaltic pump for fast delivery and removal of reagents
- One 10-port valve for multiple reagent protocols and hazardous waste segregation
- One rhomboid-shaped tray with smooth and even rocking for rapid fluid mixing and coverage
- Nine preprogrammed and four user-defined protocols for proteins and nucleic acids
- Protocol entry and editing, up to 30 steps
- Removable protocol key for storing and protects critical protocols
Specifications
39.2°F to 104°F | |
4°C to 40°C |