missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare nProtein A Sepharose™ 4 Fast Flow Affinity Media

Used for purification of monoclonal and polyclonal antibodies.   GE Healthcare Lifescience™ nProtein A Sepharose™ 4 Fast Flow Affinity Media contains native protein A coupled to Sepharose 4 Fast Flow.

Brand:  GE Healthcare 17-5280-02

Code : GX

Additional Details : Weight : 1.33810kg

 View more versions of this product

Product Code. 11369931

  • £3620.65 / 200mL


Estimated Shipment
09-02-2018
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

  • Low leakage of protein A with nearly twice total IgG binding capacity of Protein A Sepharose CL-4B
  • Used in large-scale FDA-approved processes
  • Manufactured without using animal-derived components
  • Rigidity and high degree of substitution of Sepharose 4 Fast Flow matrix facilitates speedy process of large volumes dilute cell culture fluid
  • High chemical, thermal and mechanical stability
  • Withstands high concentrations of hydrogen bond disrupting agents
Specifications

Specifications

>35mg Human IgG/mL
For the purifiCatIon of monoclonal and polyclonal antibodies at both laboratory and process scale
Native Protein A (S. aureus)
100kPa
No significant change in chromatographic performance after 1 week storage using 8M Urea, 6M Gua-HCI, 2% Benzyl Alcohol or 20% Ethanol
2 to 8°C
nProtein A Sepharose 4 fast flow media
150 to 250cm/Hour
25cm
4% Cross-Linked Agarose
200mL
20% Ethanol
90μm
2 to 10
Videos