missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare NovaSpec™ Plus Visible Spectrophotometer

Incorporates diode array technology for rapid wavelength scanning in a highly reliable and low-maintenance instrument suitable for general biotech laboratory use

Brand:  GE Healthcare 80-2117-51

Code : GE

Additional Details : Weight : 3,718.00000kg

 View more versions of this product

Product Code. 10670216

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Visible
23cm
90/265V 50/60Hz
13cm
Grafico PC utility software, serial cable, dust cover
7nm
±2nm
1.75kg
CE 89/336/EEC (EMC directive); CE 73/23/EEC (LV directive); EN-61010-1 (IEC1010-1)
Diode array
Basic enzyme kinetics, bradford, BCA, biuret and lowry protein quantitation
50/60Hz
Tungsten Halogen
90/265V
330 to 800nm
31cm
Videos