Learn More
Cytiva IMAC Sepharose™ 6 Fast Flow Affinity Media
Used for purifying proteins and peptides that have an affinity for metal ions. Cytiva IMAC Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix.
Brand: Cytiva 17-0921-07
Code : GE
Additional Details : CAS Number : 64175 Weight : 0.08400kg
Description
- Uncharged IMAC resin allows possibility to charge with metal ion of choice for optimized selectivity
- Used for ions such as histidine-tagged proteins but also untagged recombinant or native proteins
- High protein binding capacity
- Coupling chemistry and novel ligand designed to give minimal ligand leakage
- Available HiTrap™ and HiPrep™ pre-packed formats for easy scale-up and process development
Specifications
25 mL | |
For Purifying Proteins and Peptides That Have an Affinity for Metal Ions | |
IMAC Sepharose 6 Fast Flow Media | |
90 μm |
Safety and Handling
- IMAC Sepharose 6 Fast Flow, 25 mL
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol