Learn More
Cytiva Solution dNTPs
Use dNTPs incorporating the most recent advances in manufacturing technology. Cytiva illustra™ Solution dNTPs offer the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.
Brand: Cytiva 28-4065-33
Code : GH
Additional Details : Weight : 0.01400kg
Description
- Free from DNase, RNase, and nicking enzyme activity
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer-free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR and sequencing
Specifications
Liquid | |
dTTP | |
>99% | |
Colorless |
7 | |
100 mM | |
500 μm | |
Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling |