missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare illustra™ Solution dNTPs

Use dNTPs incorporating the most recent advances in manufacturing technology. GE Healthcare illustra™ Solution dNTPs offer the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.

Brand:  GE Healthcare 28-4065-23

Code : GH

Additional Details : Weight : 0.02200kg

 View more versions of this product

Product Code. 10526875

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

 

  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

 

Specifications

Specifications

100mM
Liquid
7
>99%
Colorless
Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
dGTP
500μm
Videos