Learn More
GE Healthcare illustra™ Ready-To-Go™ GenomiPhi HY DNA Amplification Kit
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60++g yield from nanogram amounts of DNA sample.
Brand:  GE Healthcare 25-6603-97
Code : GH
Additional Details : Weight : 0.48000kg
 View more versions of this product
Product Code. 12907170
  • £4537.69 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit is part of the Phi29 DNA polymerase family of products from GE Healthcare.
  • Representative amplification of the whole genome.
  • Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
  • Quick and simple automation-friendly protocol; no thermal cycler required.
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint.
  • Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
Specifications
Specifications
Midiscale whole genome amplification by isothermal strand displacement amplification
5 x 2.6mL 2X Denaturation Buffer, 5 x 96-well strips Ready-To-Go™ GenomiPhi HY cakes, 1 x vial control DNA, 12 strips of 8 domed caps
480 Reactions
Videos