Learn More
GE Healthcare illustra™ ExoProStar™ S PCR and Sequence Reaction Clean-up Kit
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
Brand:  GE Healthcare US79010
Code : GH
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15209764
  • £49.46 / 100 reactions
Estimated Shipment
06-06-2018
Add to basket
Description
Description
  • Easy to automate
  • Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
  • Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Complete heat inactivation of the enzymes within 10 minutes
  • Fast 15 minute protocol
  • Updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
  • Improves digestion efficiency with no degradation of the target PCR product
  • Sample is completely intact and ready for immediate use in manual or automated processes
Specifications
Specifications
PCR and Sequence Reaction;Removes unincorporated primers and dNTPs; Improves digestion efficiency; No loss of PCR product
PCR and Sequence Reaction Cleanup kit
100 Reactions
Videos
Special Offers
Special Offers
  • GE and Fisher Scientific brand join forces to deliver tools for Life Science applications.
    : 13007
    2018-06-30