missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for HE 33 Mini Submarine Unit

Accessory parts for HE 33 Mini Submarine

Brand:  Hoefer™ HE31A-8-1.5

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 12034737

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

0.15cm
Backless Comb
8
0.65cm
0.5cm
HE33 mini submarine unit
48.5μL
Videos
Documentation

Technical Documents

View accessory products