missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare HiTrap™ Desalting Prepacked Columns

For rapid convenient desalting and buffer exchange. HiTrap™ Desalting Prepacked Columns are prepacked, ready to use column for group separation between high and low molecular weight substances.

Brand:  GE Healthcare 11-0003-29

Code : GX

Additional Details : Weight : 1,983.00000kg

 View more versions of this product

Product Code. 11840073

  • £2816.69 / Pack of 100


Call for latest availability
+44 (0)1509 555 500
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

  • Prepacked with Sephadex™ G-25 Superfine media
  • Column material: polypropylene
  • Bed volume: 5mL
  • Bed dimensions: 16 x 25mm
  • Sample volume: 1.5mL
  • Flow velocity: <15mL/min.
  • Maximum pressure: 3 bar

Media
  • Matrix: cross-linked dextran
  • Partical size: >20μm
  • pH stability working range: 2 to 13

Specifications

Specifications

100 Pack
>20μm (Dry), 15 to 88μm (wet)
1.6cm
5mL
No
For rapid convenient desalting and buffer exchange
4° to 30°C
Videos