missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Sepharose™ Prepacked Fast Flow HIC Columns
Designed for method optimization and parameter screening
Brand: Cytiva 28-9269-86
Code : GE
Additional Details : Weight : 0.08300kg
Description
- Ready-to-use columns for convenient process development optimization
- Standard aliphatic/aromatic Hydrophobic Interaction Chromatography media for capture and intermediate purification
- Bed height of 10cm designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20cm bed height
- Small bed volume of 4.7mL for fast result and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with same chromatography media using same linear fluid velocity
- Bed dimension: 7.7 × 100mm
- Matrix: 4% cross-linked agarose/6% cross-linked agarose
- pH stability working range (media): 3 to 13
- Average particle size: 90μm
- Material (column hardware and column tube): polypropylene
- Column I.D.: 7.7mm
- Pressure max.: 1.5 bar (22psi)
- Flow velocity: <240/450cm/hr.
Specifications
1M NaOH, 30% Isopropanol, 70% Ethanol, 6M Guanidine Hydrochloride, 30% Aceto nitrile, 1mM HCl | |
4.7mL | |
7.7mm | |
3.93 in. | |
<240cm/hr. | |
Octyl | |
3 to 13 | |
4% Cross-linked Agarose | |
Ion Exchange Column |
HiScreen Octyl Sepharose 4 FF column | |
0.3 in. | |
1 x 4.7mL | |
100mm | |
Polypropylene | |
1.5 bar | |
45 to 165μm | |
4° to 30°C | |
Convenient process development optimization parameter screening |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiScreen Octyl FF
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.