missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Q Sepharose™ and SP Sepharose™ High Performance IEX Columns
Designed for method optimization and parameter screening
Brand: Cytiva 28-9505-15
Code : GE
Additional Details : Weight : 2.14100kg
Description
- Ready-to-use columns for convenient process development optimization and parameter screening
- Highly cross-linked agarose matrix with an average bead size of 34μm
- Bed height of 10cm designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20cm bed height
- Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
- Bed dimension: 7.7 × 100mm
- Matrix: 6% cross-linked agarose beads
- Average particle size: 34μm
- Max. flow velocity: <150cm/hr.
- Material (column hardware and column tube): polypropylene
- Column I.D.: 7.7mm
- Flow velocity: <150cm/hr.
HiScreen Q HP
- Ionic capacity: 0.14 to 0.20mmol Cl-/mL
- Ligand: quaternary amine
- Ion exchanger type: strong anion exchanger
- Ionic capacity: 0.15 to 0.20mmol Cl-/mL
- Ligand: sulphopropyl
- Ion exchanger type: strong cation exchanger
Specifications
8M Urea, 6M Guanidine HCl, 70% Ethanol, 1M NaOH, 1M Acetic Acid, 30% Isopropanol, 30% Aceto nitrile, 2% SDS | |
4.7mL | |
7.7mm | |
3.93 in. | |
<150cm/hr. | |
Sulphopropyl | |
4 to 13 | |
6% Cross-linked Agarose | |
Ion Exchange Column |
HiScreen SP Sepharose HP column | |
0.3 in. | |
1 x 4.7mL | |
100mm | |
Polypropylene | |
3 bar | |
45 to 165μm | |
4° to 30°C | |
Optimization and parameter screening, ion exchange chromato graphy |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiScreen SP HP
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.