Learn More
Cytiva HiScreen™ Capto™ S, Capto™ Q and Capto™ DEAE Prepacked IEX Columns
Designed for method optimization and parameter screening
Brand: Cytiva 28-9269-79
Code : GE
Additional Details : Weight : 0.08300kg
Description
- Fast, efficient, and cost-effective capture and intermediate protein purification
- Ready-to-use columns for convenient process development optimization
- Highly cross-linked agarose with dextran surface extender matrix
- Bed height of 10cm is designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20cm bed height
- Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
- Combines high capacity with high flow velocity and low backpressure to reduce process cycle times and increase productivity
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
Capto S: Strong cation (sulfonate group) exchange BioProcess™ medium
Capto Q: Strong anion (quartenary amine group) exchange BioProcess medium
Capto DEAE: Weak anion (diethylaminoethyl group) exchange BioProcess medium
Specifications
1M Acetic Acid, 1M NaOH, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol | |
4.7mL | |
7.7mm | |
3.93 in. | |
<700cm/hr. | |
Sulphonate group | |
4 to 12 | |
Highly cross-linked Agarose with Dextran surface extender | |
Ion Exchange Column |
HiScreen Capto S IEX column | |
0.3 in. | |
1 x 4.7mL | |
100mm | |
Polypropylene | |
3 bar | |
45 to 165μm | |
4° to 30°C | |
Modern large-scale manufacturers for fast, efficient and cost-effective capture and intermediate Protein purification, optimization and parameter screening |
Safety and Handling
- HiScreen Capto S
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-18.5 per cent of the mixture consists of component(s) of unknown acute toxicity.