missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare His MultiTrap™ 96-well Filter Plates

Prepacked multiwell filter plates for high reproducibility in expression screening and small-scale purification of histidine-tagged proteins

Brand:  GE Healthcare 28-4009-89

Code : GE

Additional Details : Weight : 0.52160kg


 View more versions of this product

Product Code. 11713319

  • £557.52 / Pack of 4


Estimated Shipment
08-06-2018
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

  • No filtration needed: load unclarified sample directly, increasing reproducibility and saving time
  • Compatible with robotic systems or manual protocols with centrifuge or vacuum
  • Easy scale-up
  • Long-term pH range 3 to 12

  • Protein binding capacity up to 800μg histidine-tagged protein/well with His MultiTrap FF
  • Protein binding capacity up to 1mg histidine-tagged protein/well with His MultiTrap HP

Specifications

Specifications

Polypropylene, Polyethylene
34μm
4 x 96 Well Plate
6% Highly Cross-linked Agarose
3 to 12
His MultiTrap HP Filter Plates
4° to 30°C
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .