Learn More
GE Healthcare High Purity Solution ddNTPs
Use in any application that requires ddNTPs of high purity. GE Healthcare Life Sciences™ High Purity Solution ddNTPs are available either as individual products or in a convenient set.
Brand:  GE Healthcare 27-2051-01
Code : GH
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 10544195
  • £146.15 / Each
Estimated Shipment
11-06-2018
Add to basket
If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.
Description
Description
  • Each lot of ddNTPs is functionally tested to meet or exceed criteria for high-quality sequencing with Thermo Sequenase™ DNA polymerase
  • High purity: >98% triphosphate
Dideoxy sequencing, Genotyping reactions
Specifications
Specifications
100mM
Liquid
ddATP
4μmol (40μL)
Colorless
Dideoxy sequencing, genotyping reactions
≥98%
Videos
Documentation
Technical Documents