missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare HE 33 Combs

Without back

Brand:  GE Healthcare 80605701

Code : GH

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 11370462

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

0.3cm
Backless Comb
20
0.47cm
HE99X max submarine electrophoresis unit
Videos