missing translation for 'onlineSavingsMsg'
Learn More

Biochrom™ GeneQuant™ 1300 Spectrophotometers

Can be used with a wide variety of cuvettes or capillaries for sample volumes from 3μL to 2mL

Brand:  Biochrom™ 80-2120-02

Code : GE

Additional Details : Weight : 5.28000kg

 View more versions of this product

Product Code. 10574815

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

CCD array
DNA, RNA and oligonucleotide purity and concentration measurements, quantitation of proteins, nucleic acids and bacterial cell cultures, cell density and enzyme kinetics, cDNA application software for measuring incorporation of CyTM3 and Cy5 or other dyes into purified microarray hybridization probes and PCR products
50/60Hz
5nm
±2nm
±0.5nm
10cm
GeneQuant 1300 Spectrophotometer with Bluetooth
100 to 240V 50/60Hz
39cm
Xenon
100 to 240V
190 to 1100nm
4.5kg
3 Year
Videos