missing translation for 'onlineSavingsMsg'
Learn More

Biochrom™ GeneQuant™ 1300 Spectrophotometers

Can be used with a wide variety of cuvettes or capillaries for sample volumes from 3μL to 2mL

Brand:  Biochrom™ 80-2120-00

Code : GE

Additional Details : Weight : 5.45400kg

 View more versions of this product

Product Code. 10564815

  • £4998.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Specifications

Specifications

CCD array
DNA, RNA and oligonucleotide purity/concentration measurements
50/60Hz
5nm
±2nm
±0.5nm
10cm
GeneQuant 1300 Spectrophotometer
100 to 240V 50/60Hz
39cm
Xenon
100/240V
190 to 1100nm
4.5kg
3 Year
Videos