missing translation for 'onlineSavingsMsg'
Learn More

Biochrom™ GeneQuant™ 1300 Spectrophotometers

Can be used with a wide variety of cuvettes or capillaries for sample volumes from 3μL to 2mL

Brand:  Biochrom™ 80-2120-01

Code : GE

Additional Details : Weight : 5.67900kg

 View more versions of this product

Product Code. 10034834

  • £5815.00 / Each

Estimated Shipment
12-04-2021

Please to purchase this item. Need a web account? Register with us today!


If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

The GeneQuant 1300 spectrophotometer is a compact instrument pre- programmed with a range of methods for the quantitation of proteins, nucleic acids, and bacterial cell cultures. Absorbance and concentration measurements can be made at any wavelength, providing flexibility for future applications.

  • Built-in applications for cell density, nucleic acids, proteins, and enzyme kinetics
  • cDNA application software for measuring incorporation of Cy™3 and Cy™5, or other dyes into purified microarray hybridization probes and PCR products
  • Wavelength range of 190 to 1100nm, with wavelength scan from 200 to 900nm performed in less than 5 sec. with zoom facility, peak identification, and on-peak confirmation
  • Visual inspection of a nucleic acid scan can identify the presence of impurities especially useful with RNA samples
  • Storage for 90 user-defined methods with (optional) password protection
  • Large, easy-to-read graphical display
  • Optics with no moving parts for high-energy throughput combined with a long-life Xenon lamp
  • Optional built-in printer or output data via USB to a PC for printing or storage
  • Wireless bluetooth connectivity (optional)

Specifications

Specifications

CCD array
100 to 240V 50/60Hz
39cm
Xenon
100 to 240V
±0.5nm
10cm
GeneQuant 1300 Spectrophotometer with printer
190 to 1,100nm
DNA, RNA and oligonucleotide purity and concentration measurements, quantitation of proteins, nucleic acids and bacterial cell cultures, cell density and enzyme kinetics, cDNA application software for measuring incorporation of CyTM3 and Cy5 or other dyes into purified microarray hybridization probes and PCR products
50/60Hz
5nm
190 to 1100nm
4.5kg
3 Year
Videos
SDS
Documents