missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GammaBind™ G Sepharose™ Medium
Use recombinant GammaBind™ for laboratory scale antibody purification. Cytiva Life Sciences™ GammaBind™ G Sepharose™ Medium binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
£644.00 - £6501.62
Specifications
Product Type | Sepharose Medium |
---|---|
Binding Capacity | >18 mg/mL Human IgG, 6 mg/mL Mouse IgG |
Ligand Concentration | ≈3 mg GammaBind G, Type 2/mL drained gel |
Particle Size | 45 to 165 μm |
pH | 3 to 9 |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11574945
|
Cytiva
17-0885-04 |
250 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11554945
|
Cytiva
17-0885-01 |
5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11564945
|
Cytiva
17-0885-02 |
25 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Binds to all IgG subclasses from human, mouse, and rat and total IgG from guinea pig, rabbit, goat, cow, sheep, and horse. Engineered for antibody purification at laboratory scale.
- Will not cross-react with other serum proteins such as IgM, IgE, IgA or transferrin
- Ligand: GammaBind G type 2 lacking albumin-binding region
- Ligand coupling method: cyanogen bromide activation
- Ligand density: ≈ 3mg GammaBind G, type 2/mL medium
- Number of IgG binding sites per ligand: 2
- Matrix: Agarose, 4%
- Average particle size: 90μm
- Binding capacity: >18 mg human IgG/ml, 6 mg mouse IgG/ml
- pH stability: 3-9 (working), 2-9 (short term)
Specifications
Sepharose Medium | |
≈3 mg GammaBind G, Type 2/mL drained gel | |
3 to 9 | |
PBS pH 7, 20% Ethanol |
>18 mg/mL Human IgG, 6 mg/mL Mouse IgG | |
45 to 165 μm | |
For protein purification by affinity coupling |