missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Filler Sheet

1mm Ettan DALT filter sheets

Brand:  Cytiva 80-6467-60

Code : A0

Additional Details : Weight : 0.50200kg

Product Code. 10667525

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • pack size: 6
  • 1mm Sheet thickness

Specifications

Specifications

Filler Sheet
6 Pack
0.03 in.
0.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers