Learn More
GE Healthcare Dithiothreitol (DTT)
Include reducing agents in the sample solution to break any disulfide bonds present and to maintain all proteins in their fully reduced state. GE Healthcare Life Sciences™ Dithiothreitol (DTT) is one of the most commonly used reductants.
Brand:  GE Healthcare 17-1318-01
Code : GE
Additional Details : CAS Number : 3483-12-3 Weight : 0.01800kg
 View more versions of this product
Product Code. 10666885
  • £36.69 / 1g
Estimated Shipment
08-02-2018
Add to basket
If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.
Description
Description
  • Use for IEF, SDS-PAGE
  • White
  • Solid
  • Easily soluble in cold water and hot water
  • Threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT
Specifications
Specifications
IEF, SDS-PAGE
0 to 5.1
1000mg
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to A§.