GE Healthcare Dharmacon TOX™ Transfection Control
Quick and easy to use, requiring only visual inspection of cells
Overview
Brand: GE Healthcare Dharmacon
Manufacturer Part Number: D-001500-01-05
UNSPSC: 41106402
Code: P1
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11597200
Quantity Price
1 £ 139.12 / 5nmol
Estimated Shipment date
from Supplier 03-04-2017
Add to basket
Description and Specification
Specification
Product Type siCONTROL TOX Transfection Control
For Use With (Application) Ideal for optimizing transfection condition as it provide an easily quantifiable measure of cellular uptake leading to silencing
Quantity 5nmol